missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD1B Antibody [HRP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37990H
This item is not returnable.
View return policy
Description
JMJD1B Polyclonal antibody specifically detects JMJD1B in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| JMJD1B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| HRP | |
| C5orf7, EC 1.14.11, EC 1.14.11.-, JHDM2B, JmjC domain-containing histone demethylation protein 2B, JMJD1B, jumonji domain containing 1B, Jumonji domain-containing protein 1B, KIAA1082chromosome 5 open reading frame 7, lysine (K)-specific demethylase 3B, lysine-specific demethylase 3B, NET22,5qNCA, Nuclear protein 5qNCA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-600 of human JMJD1B (NP_057688.2).,, Sequence:, GAENKEAGKTLEQVGQGIVASAAVVTTASSTPNTVRISDTGLAAGTVPEKQKGSRSQASGENSRNSILASSGFGAPLPSSSQPLTFGSGRSQSNGVLATENKPLGFSFGCSSAQEAQKDTDLSKNLFFQCMSQTLPTSNYFTTVSESLADDSSSRDSFKQSLESLSSGLCKGRSVLGTDTKPGSKAGSSVDRKVPAESMPT | |
| 0.1 mL | |
| Cancer | |
| 51780 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction