missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF3 Antibody [HRP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35704H
This item is not returnable.
View return policy
Description
DPF3 Polyclonal antibody specifically detects DPF3 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DPF3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8110 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| HRP | |
| CERD4, D4, zinc and double PHD fingers, family 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-200 of human DPF3 (NP_036206.3).,, Sequence:, DPKLRLLEIKPEVELPLKKDGFTSESTTLEALLRGEGVEKKVDAREEESIQEIQRVLENDENVEEGNEEEDLEEDIPKRKNRTRGRARGSAGGRRRHDAASQEDHDKPYVC | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction