missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody [PE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38070PE
This item is not returnable.
View return policy
Description
Peripherin Polyclonal antibody specifically detects Peripherin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Peripherin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PE | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human Peripherin (NP_006253.2).,, Sequence:, MSHHPSGLRAGFSSTSYRRTFGPPPSLSPGAFSYSSSSRFSSSRLLGSASPSSSVRLGSFRSPRAGAGALLRLPSERLDFSMAEALNQEFLATRSNEKQELQELNDRFANFIEKVRFLEQQNAALRGELSQARGQEPARADQLCQQELRELRRELELLGRERDRVQVERDGLAEDLAALKQRLEEETRKREDAEHNLVLFRKDVDDATLSRLELERKIESLMDEIEFLKKLHEEELRDLQVSVESQQVQQVEVEATVKPE | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 5630 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction