missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suppressor of Fused Antibody [FITC], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35342F
This item is not returnable.
View return policy
Description
Suppressor of Fused Polyclonal antibody specifically detects Suppressor of Fused in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Suppressor of Fused | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| FITC | |
| SUFUHSUFUXLPRO1280, suppressor of fused homolog, suppressor of fused homolog (Drosophila) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 325-484 of human Suppressor of Fused (NP_057253.2).,, Sequence:, PPINPQRQNGLAHDRAPSRKDSLESDSSTAIIPHELIRTRQLESVHLKFNQESGALIPLCLRGRLLHGRHFTYKSITGDMAITFVSTGVEGAFATEEHPYAAHGPWLQILLTEEFVEKMLEDLEDLTSPEEFKLPKEYSWPEKKLKVSILPDVVFDSPLH | |
| 0.1 mL | |
| Cancer | |
| 51684 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction