missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIC8A Antibody [Janelia Fluor« 525], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35138JF525
This item is not returnable.
View return policy
Description
RIC8A Polyclonal antibody specifically detects RIC8A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RIC8A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 60626 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| Janelia Fluor 525 | |
| RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 362-531 of human RIC8A (NP_001273063.1).,, Sequence:, DVRTRPEVGEMLRNKLVRLMTHLDTDVKRVAAEFLFVLCSESVPRFIKYTGYGNAAGLLAARGLMAGGRPEGQYSEDEDTDTDEYKEAKASINPVTGRVEEKPPNPMEGMTEEQKEHEAMKLVTMFDKLSRNRVIQPMGMSPRGHLTSLQDAMCETMEQQLSSDPDSDPD | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction