missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabpb2 Antibody [DyLight 680], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38042FR
This item is not returnable.
View return policy
Description
Gabpb2 Polyclonal antibody specifically detects Gabpb2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Gabpb2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 126626 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| DyLight 680 | |
| GA binding protein transcription factor, beta subunit 2, GA-binding protein subunit beta-2, GABP subunit beta-2, GABPB-2, MGC29891 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 239-448 of human Gabpb2 (NP_653219.1).,, Sequence:, SNSHRATANTEEIIEGNSVDSSIQQVMGSGGQRVITIVTDGVPLGNIQTSIPTGGIGQPFIVTVQDGQQVLTVPAGKVAEETVIKEEEEEKLPLTKKPRIGEKTNSVEESKEGNERELLQQQLQEANRRAQEYRHQLLKKEQEAEQYRLKLEAIARQQPNGVDFTMVEEVAEVDAVVVTEGELEERETKVTGSAGTTEPHTRVSMATVSS | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction