missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUG1 Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35540AF594
This item is not returnable.
View return policy
Description
SUG1 Polyclonal antibody specifically detects SUG1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| SUG1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5705 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 594 | |
| MSUG1 protein, p45, p45/SUG26S protease regulatory subunit 8, proteasome (prosome, macropain) 26S subunit, ATPase, 5, proteasome 26S ATPase subunit 5, Proteasome 26S subunit ATPase 5, Proteasome subunit p45, S8, SUG-1, SUG126S proteasome AAA-ATPase subunit RPT6, Tat-binding protein homolog 10, TBP10, thyroid receptor interactor 1, TRIP1Thyroid hormone receptor-interacting protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 127-406 of human SUG1 (NP_002796.4).,, Sequence:, LPNKVDPLVSLMMVEKVPDSTYEMIGGLDKQIKEIKEVIELPVKHPELFEALGIAQPKGVLLYGPPGTGKTLLARAVAHHTDCTFIRVSGSELVQKFIGEGARMVRELFVMAREHAPSIIFMDEIDSIGSSRLEGGSGGDSEVQRTMLELLNQLDGFEATKNIKVIMATNRIDILDSALLRPGRIDRKIEFPPPNEEARLDILKIHSRKMNLTRGINLRKIAELMPGASGAEVKGVCTEAGMYALRERRVHVTQEDFEMAVAKVMQKDSEKNMSIKKLWK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction