missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LASP1 Antibody [DyLight 594], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38061DL594
This item is not returnable.
View return policy
Description
LASP1 Polyclonal antibody specifically detects LASP1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| LASP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3927 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 594 | |
| LASP-1, LIM and SH3 domain protein 1, LIM and SH3 protein 1, Metastatic lymph node gene 50 protein, MLN 50, MLN50Lasp-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-205 of human LASP1 (NP_006139.1).,, Sequence:, RMGPSGGEGMEPERRDSQDGSSYRRPLEQQQPHHIPTSAPVYQQPQQQPVAQSYGGYKEPAAPVSIQRSAPGGGGK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction