missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCAR1 Antibody [Biotin], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35368B
This item is not returnable.
View return policy
Description
CCAR1 Polyclonal antibody specifically detects CCAR1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| CCAR1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| Biotin | |
| CARP-1, CARP1MGC44628, Cell cycle and apoptosis regulatory protein 1, cell division cycle and apoptosis regulator 1, cell division cycle and apoptosis regulator protein 1, Death inducer with SAP domain, DIS, FLJ10590, RP11-437A18.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CCAR1 (NP_060707.2).,, Sequence:, MAQFGGQKNPPWATQFTATAVSQPAALGVQQPSLLGASPTIYTQQTALAAAGLTTQTPANYQLTQTAALQQQAAAAAAALQQQYSQPQQALYSVQQQLQQPQQTLLTQPAVALPTSLSLSTPQPTAQITVSYPTPRSSQQQTQPQKQRVFTGVVTKLHDTFGFVDEDVFFQLSAVKGKTPQVGDRVLVEATYNPNMPFKW | |
| 0.1 mL | |
| Apoptosis, Cell Cycle and Replication, DNA Repair | |
| 55749 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction