missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 4/CHRNA4 Antibody [DyLight 680], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35674FR
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 4/CHRNA4 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Nicotinic Acetylcholine R alpha 4/CHRNA4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| DyLight 680 | |
| BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-242 of human Nicotinic Acetylcholine R alpha 4/CHRNA4 (P43681).,, Sequence:, HVETRAHAEERLLKKLFSGYNKWSRPVANISDVVLVRFGLSIAQLIDVDEKNQMMTTNVWVKQEWHDYKLRWDPADYENVTSIRIPSELIWRPDIVLYNNADGDFAVTHLTKAHLFHDGRVQWTPPAIYKSSCSIDVTFFPFDQQNCTMKFGSWTYDKAKIDLVNMHSRVDQLDFWESGEWVIVDAVGTYNTRKYECCAEIYPDITYAFVIRRL | |
| 0.1 mL | |
| Neuroscience | |
| 1137 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction