missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp72 Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-27898MFV500
This item is not returnable.
View return policy
Description
ERp72 Polyclonal antibody specifically detects ERp72 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ERp72 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| mFluor Violet 500 SE | |
| Endoplasmic reticulum resident protein 70, Endoplasmic reticulum resident protein 72, ER protein 70, ERp70, ERP70ER protein 72, ERp72, ERp-72, ERP72EC 5.3.4.1, intestinal-related), protein disulfide isomerase family A, member 4, protein disulfide isomerase related protein (calcium-binding protein, protein disulfide isomerase-associated 4, protein disulfide-isomerase A4 | |
| ERp72 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human ERp72. The immunogen is located within the last 50 amino acids of ERp72. | |
| 0.1 mL | |
| Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 9601 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction