missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3A Antibody [DyLight 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35076G
This item is not returnable.
View return policy
Description
EIF3A Polyclonal antibody specifically detects EIF3A in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| EIF3A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 488 | |
| centrosomin homolog, cytoplasmic protein p167, eIF3 p167, eIF3 p180, eIF3 p185, EIF3, p180 subunit, eIF3a, eIF3-p170, EIF3S10EIF3, eIF-3-theta, eIF3-theta, Eukaryotic translation initiation factor 3 subunit 10, eukaryotic translation initiation factor 3 subunit A, eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD), eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD), eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa, eukaryotic translation initiation factor 3, subunit 10, 170kD, eukaryotic translation initiation factor 3, subunit A, KIAA0139P167, p180, p185, TIF32 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human EIF3A (NP_003741.1).,, Sequence:, MPAYFQRPENALKRANEFLEVGKKQPALDVLYDVMKSKKHRTWQKIHEPIMLKYLELCVDLRKSHLAKEGLYQYKNICQQVNIKSLEDVVRAYLKMAEEKTEAAKEESQQMVLDIEDLDNIQTPESVLLSAVSGEDTQDRTDRLLLTPWVKFLWESYRQCLDLLRNNSRVERLYHDIAQQAFKFCLQYTRKAEFRKLCDNLRMHLSQIQRHHNQSTAINLNNPESQSMHLETRLVQLDSAISMELWQEAF | |
| 0.1 mL | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 8661 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction