missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scaffold attachment factor B2 Antibody [PE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38086PE
This item is not returnable.
View return policy
Description
Scaffold attachment factor B2 Polyclonal antibody specifically detects Scaffold attachment factor B2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Scaffold attachment factor B2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9667 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| PE | |
| KIAA0138SAF-B2, scaffold attachment factor B2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human Scaffold attachment factor B2 (NP_055464.1).,, Sequence:, MAETLPGSGDSGPGTASLGPGVAETGTRRLSELRVIDLRAELKKRNLDTGGNKSVLMERLKKAVKEEGQDPDEIGIELEATSKKSAKRCVKGLKMEEEGTEDNGLEDDSRDGQEDMEASLENLQNMGMMDMSVLDETEVANSSAPDFGEDGTDGLLDSFCDSKEYVAAQLRQLPAQPPEHAVDGEGFKNTLETSSLNFKVTPDIEESLLEPENEKILDILGETCKSEPVKEESSELEQPFAQDTSSVGPDRKLAE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction