missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dnmt3L Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35367MFV610
This item is not returnable.
View return policy
Description
Dnmt3L Polyclonal antibody specifically detects Dnmt3L in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Dnmt3L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| mFluor Violet 610 SE | |
| DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 151-386 of human Dnmt3L (NP_787063.1).,, Sequence:, SGLLQRRRKWRSQLKAFYDRESENPLEMFETVPVWRRQPVRVLSLFEDIKKELTSLGFLESGSDPGQLKHVVDVTDTVRKDVEEWGPFDLVYGATPPLGHTCDRPPSWYLFQFHRLLQYARPKPGSPRPFFWMFVDNLVLNKEDLDVASRFLEMEPVTIPDVHGGSLQNAVRVWSNIPAIRSRHWALVSEEELSLLAQNKQSSKLAAKWPTKLVKNCFLPLREYFKYFSTELTSSL | |
| 0.1 mL | |
| Chromatin Research | |
| 29947 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction