missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOSIP Antibody [DyLight 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35095G
This item is not returnable.
View return policy
Description
NOSIP Polyclonal antibody specifically detects NOSIP in Human samples. It is validated for ELISA,Western Blot
Specifications
| NOSIP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51070 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| DyLight 488 | |
| CGI-25, eNOS interacting protein, eNOS-interacting protein, nitric oxide synthase interacting protein, nitric oxide synthase-interacting protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human NOSIP (NP_057037.1).,, Sequence:, MTRHGKNCTAGAVYTYHEKKKDTAASGYGTQNIRLSRDAVKDFDCCCLSLQPCHDPVVTPDGYLYEREAILEYILHQKKEIARQMKAYEKQRGTRREEQKELQRAASQDHVRGFLEKESAIVSRPLNPFTAKALSGTSPDDVQPGPSVGPPSKDKDKVLPSFWIPSLTPEAKATKLEKPSRTVTCPMSGKPLRMSDLTPVHFTPLDSSVDRVGLITRSERYVCAVTRDSLSNATPCAVLRPSGAVVTLECVEKLIRKDMVDPVTGDKLTDRDIIVLQRGGTGFAGSGVKLQAEKSRPVMQA | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction