missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAWR/ PAR4 Antibody [Janelia Fluor« 525], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
PAWR / PAR4 Polyclonal antibody specifically detects PAWR / PAR4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
Tekniske data
| Antigen | PAWR / PAR4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Par-4, PAR4prostate apoptosis response protein 4, PRKC apoptosis WT1 regulator protein, PRKC, apoptosis, WT1, regulator, Prostate apoptosis response 4 protein, prostate apoptosis response protein PAR-4, transcriptional repressor PAR4, WT1-interacting protein |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200-300 of human PAWR/ PAR4 (NP_002574.2),, Sequence:, QNEAVNLLDPGSSYLLQEPPRTVSGRYKSTTSVSEEDVSSRYSRTDRSGFPRYNRDANVSGTLVSSSTLEKKIEDLEKEVVRERQENLRLVRLMQDKEEMI |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?