missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACYBP Antibody (7D11A8) [Alexa Fluor« 750], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
CACYBP Monoclonal antibody specifically detects CACYBP in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Serum
Specifications
Specifications
| Antigen | CACYBP |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 7D11A8 |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CacyBP, calcyclin binding protein, GIG5, hCacyBP, MGC87971, RP1-102G20.6, S100A6-binding protein, S100A6BPgrowth-inhibiting gene 5 protein, Siah-interacting protein, Siah-interacting protein (SIP), SIPcalcyclin-binding protein |
| Host Species | Mouse |
| Immunogen | Truncated recombinant CACYBP expressed in E. Coli strain BL21 (DE3). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?