missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FANCI Polyclonal antibody specifically detects FANCI in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | FANCI |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1061-1328 of human FANCI (NP_001106849.1).,, Sequence:, DQDVEVEKTNHFAIVNLRTAAPTVCLLVLSQAEKVLEEVDWLITKLKGQVSQETLSEEASSQATLPNQPVEKAIIMQLGTLLTFFHELVQTALPSGSCVDTLLKDLCKMYTTLTALVRYYLQVCQSSGGIPKNMEKLVKLSGSHLTPLCYSFISYVQNKSKSLNYTGEKKEKPAAVATAMARVLRETKPIPNLIFAIEQYEKFLIHLSKKSKVNLMQHMKLSTSRDFKIKGNILDMVLREDGEDENEEGTASEHGGQNKEPAKKKRKK |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?