missing translation for 'onlineSavingsMsg'
Learn More

PEX10 Antibody [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30498140 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498140 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498140 Supplier Novus Biologicals Supplier No. NBP335707G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PEX10 Polyclonal antibody specifically detects PEX10 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-160 of human PEX10 (NP_002608.1).,, Sequence:, EVIRAAQKDEYYRGGLRSAAGGALHSLAGARKWLEWRKEVELLSDVAYFGLTTLAGYQTLGEEYVSIIQVDPSRIHVPSSLRRGVLVTLHAVLPYLLDKALLPLEQELQADPDSGRPLQGSLGPGGRGCSGARRWMRHHTATLTEQQRRAL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 5192
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.