missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NUP98 Polyclonal antibody specifically detects NUP98 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | NUP98 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 621-920 of human NUP98 (NP_624357.1).,, Sequence:, SKPVDENHQQDGDEDSLVSHFYTNPIAKPIPQTPESAGNKHSNSNSVDDTIVALNMRAALRNGLEGSSEETSFHDESLQDDREEIENNSYHMHPAGIILTKVGYYTIPSMDDLAKITNEKGECIVSDFTIGRKGYGSIYFEGDVNLTNLNLDDIVHIRRKEVVVYLDDNQKPPVGEGLNRKAEVTLDGVWPTDKTSRCLIKSPDRLADINYEGRLEAVSRKQGAQFKEYRPETGSWVFKVSHFSKYGLQDSDEEEEEHPSKTSTKKLKTAPLPPASQTTPLQMALNGKPAPPPQVEKKGQ |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?