missing translation for 'onlineSavingsMsg'
Learn More

TCP1 alpha Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30498054 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498054 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498054 Supplier Novus Biologicals Supplier No. NBP335337PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCP1 alpha Polyclonal antibody specifically detects TCP1 alpha in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP1 alpha
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 307-556 of human TCP1 alpha (NP_110379.2).,, Sequence:, AVRRVLKRDLKRIAKASGATILSTLANLEGEETFEAAMLGQAEEVVQERICDDELILIKNTKARTSASIILRGANDFMCDEMERSLHDALCVVKRVLESKSVVPGGGAVEAALSIYLENYATSMGSREQLAIAEFARSLLVIPNTLAVNAAQDSTDLVAKLRAFHNEAQVNPERKNLKWIGLDLSNGKPRDNKQAGVFEPTIVKVKSLKFATEAAITILRIDDLIKLHPESKDDKHGSYEDAVHSGALND
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6950
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.