missing translation for 'onlineSavingsMsg'
Learn More

NUDT21 Antibody [PE], Novus Biologicals Biologicals™

Product Code. 30498049 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498049 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498049 Supplier Novus Biologicals Supplier No. NBP338098PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NUDT21 Polyclonal antibody specifically detects NUDT21 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT21
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias CFIm25, CFIM25CPSF 25 kDa subunit, cleavage and polyadenylation specific factor 5, 25 kD subunit, cleavage and polyadenylation specific factor 5, 25 kDa, Cleavage and polyadenylation specificity factor 25 kDa subunit, cleavage and polyadenylation specificity factor subunit 5, CPSF25, CPSF5DKFZp686H1588, Nucleoside diphosphate-linked moiety X motif 21, nudix (nucleoside diphosphate linked moiety X)-type motif 21, Nudix motif 21, pre-mRNA cleavage factor Im (25kD), Pre-mRNA cleavage factor Im 25 kDa subunit, pre-mRNA cleavage factor Im, 25kD subunit
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human NUDT21 (NP_008937.1).,, Sequence:, MSVVPPNRSQTGWPRGVTQFGNKYIQQTKPLTLERTINLYPLTNYTFGTKEPLYEKDSSVAARFQRMREEFDKIGMRRTVEGVLIVHEHRLPHVLLLQLGTTFFKLPGGELNPGEDEVEGLKRLMTEILGRQDGVLQDWVIDDCIGNWWRPNFEPPQYPYIPAHITKPKEHKKLFLVQLQEKALFAVPKNYKLVAAPLFELYDNAPGYGPIISSLPQLLSRFNFIYN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 11051
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.