missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRFS1 Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
UQCRFS1 Polyclonal antibody specifically detects UQCRFS1 in Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | UQCRFS1 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Complex III subunit 5, Cytochrome b-c1 complex subunit 5, cytochrome b-c1 complex subunit Rieske, mitochondrial, EC 1.10.2.2, Rieske iron-sulfur protein, RIP1, RIS1, RISPUbiquinol-cytochrome c reductase iron-sulfur subunit, ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1, UQCR5 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 79-274 of human UQCRFS1 (NP_005994.2).,, Sequence:, SHTDIKVPDFSEYRRLEVLDSTKSSRESSEARKGFSYLVTGVTTVGVAYAAKNAVTQFVSSMSASADVLALAKIEIKLSDIPEGKNMAFKWRGKPLFVRHRTQKEIEQEAAVELSQLRDPQHDLDRVKKPEWVILIGVCTHLGCVPIANAGDFGGYYCPCHGSHYDASGRIRLGPAPLNLEVPTYEFTSDDMVIVG |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?