missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UGT1A9 Polyclonal antibody specifically detects UGT1A9 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | UGT1A9 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.4.1.17, GNT1, HLUGP4, LUGP4, polypeptide A9, UDP glucuronosyltransferase 1 family, polypeptide A9, UDP-glucuronosyltransferase 1-9, UDP-glucuronosyltransferase 1A9, UDP-glucuronosyltransferase 1-I, UDPGT, UDPGT 1-9, UGT1, UGT1*9, UGT1.9, UGT1-09, UGT1-9, UGT1AI, UGT1I, UGT-1I |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 26-250 of human UGT1A9 (NP_066307.1).,, Sequence:, GKLLVVPMDGSHWFTMRSVVEKLILRGHEVVVVMPEVSWQLGRSLNCTVKTYSTSYTLEDLDREFKAFAHAQWKAQVRSIYSLLMGSYNDIFDLFFSNCRSLFKDKKLVEYLKESSFDAVFLDPFDNCGLIVAKYFSLPSVVFARGILCHYLEEGAQCPAPLSYVPRILLGFSDAMTFKERVRNHIMHLEEHLLCHRFFKNALEIASEILQTPVTEYDLYSHTSI |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?