missing translation for 'onlineSavingsMsg'
Learn More

[3-(Trifluoromethyl)phenylthio]acetic acid, Thermo Scientific™

Catalog Number p-7033933
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
349-83-7
C9H7F3O2S
236.208
MFCD00052114
YBUZSRSMQIEFBR-UHFFFAOYSA-N
2801046
2-[3-(trifluoromethyl)phenyl]sulfanylacetic acid
C1=CC(=CC(=C1)SCC(=O)O)C(F)(F)F
This item is not returnable. View return policy
349-83-7
C9H7F3O2S
236.208
MFCD00052114
YBUZSRSMQIEFBR-UHFFFAOYSA-N
2801046
2-[3-(trifluoromethyl)phenyl]sulfanylacetic acid
C1=CC(=CC(=C1)SCC(=O)O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 236.21
Chemical Name or Material [3-(Trifluoromethyl)phenylthio]acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'