missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiophenemethylamine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store in cool, dry place in tightly closed container. With good ventilation. Store away from air.
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store in cool, dry place in tightly closed container. With good ventilation. Store away from air.
Specifications
Density | 1.13 |
Boiling Point | 102°C (15 mmHg) |
Refractive Index | 1.566 |
Quantity | 1 g |
UN Number | UN2735 |
Sensitivity | Air sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 113.18 |
Percent Purity | 97% |
Chemical Name or Material | 3-Thiophenemethylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'