missing translation for 'onlineSavingsMsg'
Learn More

3'-(Pentafluorothio)acetophenone, 97%, Thermo Scientific™

Catalog Number p-7052445
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
159896-31-8
C8H7F5OS
246.20
MFCD16652407
GUYKQTMZBIAURU-UHFFFAOYSA-N
3'-(Pentafluorosulfur)acetophenone
1-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]ethan-1-one
CC(=O)C1=CC(=CC=C1)S(F)(F)(F)(F)F
This item is not returnable. View return policy
159896-31-8
C8H7F5OS
246.20
MFCD16652407
GUYKQTMZBIAURU-UHFFFAOYSA-N
3'-(Pentafluorosulfur)acetophenone
1-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]ethan-1-one
CC(=O)C1=CC(=CC=C1)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 43°C to 45°C
Color White
Quantity 1 g
Formula Weight 246.19
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 3'-(Pentafluorothio)acetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'