missing translation for 'onlineSavingsMsg'
Learn More

3-O-Benzyl-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose, 97%, Thermo Scientific™

Catalog Number p-7043202
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18685-18-2
C19H26O6
350.411
MFCD00269929
ZHFVGOMEUGAIJX-QTPCEWDRSA-N
3-o-benzyl-1,2:5,6-di-o-isopropylidene-, a-d-glucofuranose, 1,2:5,6-bis-o-1-methylethylidene-3-o-phenylmethyl-, 4-3ar,5r,6as-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl-2,2-dimethyl-1,3-dioxolane
45038272
(3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)OCC4=CC=CC=C4)C
This item is not returnable. View return policy
18685-18-2
C19H26O6
350.411
MFCD00269929
ZHFVGOMEUGAIJX-QTPCEWDRSA-N
3-o-benzyl-1,2:5,6-di-o-isopropylidene-, a-d-glucofuranose, 1,2:5,6-bis-o-1-methylethylidene-3-o-phenylmethyl-, 4-3ar,5r,6as-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl-2,2-dimethyl-1,3-dioxolane
45038272
(3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)OCC4=CC=CC=C4)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A derivative of glucofuranose.

Solubility
Soluble in chloroform

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.094
Boiling Point 140°C to 142°C
Refractive Index 1.4925
Quantity 1 g
Solubility Information Soluble in chloroform
Optical Rotation −30° (c=1 in Ethanol)
Formula Weight 350.41
Percent Purity 97%
Physical Form Oil
Chemical Name or Material 3-O-Benzyl-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'