Learn More
3-Methylbutanol, Puriss. p.a., ACS Reagent, ≥98.5% (GC), Honeywell Riedel-de Haën™
Catalog Number p-7120088
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
123-51-3
C5H12O
88.15
MFCD00002934
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO
123-51-3
C5H12O
88.15
MFCD00002934
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO
Puriss. p.a., ACS Reagent,≥98.5% (GC)
ACS GENERAL USE SOLVENTS
  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.809g/mL at 20°C
Boiling Point 131°C to 132°C
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2OH
Refractive Index 1.407
Quantity 250 mL
UN Number UN1105
Beilstein 1718835
Formula Weight 88.15g/mol
Percent Purity ≥98.5% (GC)
Chemical Name or Material 3-Methylbutanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methylbutanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.