missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid, 97%, Thermo Scientific™

Catalog Number p-4602057
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
603122-41-4
C9H11BO5
210
YCXPWNGIPLGCOJ-UHFFFAOYSA-N
3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, ycxpwngiplgcoj-uhfffaoysa-n, methyl 4-borono-2-methoxybenzoate
17750256
This item is not returnable. View return policy
603122-41-4
C9H11BO5
210
YCXPWNGIPLGCOJ-UHFFFAOYSA-N
3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, ycxpwngiplgcoj-uhfffaoysa-n, methyl 4-borono-2-methoxybenzoate
17750256
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 210
Percent Purity 97%
Chemical Name or Material 3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
compliance-icons
Product Identifier
  • 3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'