missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034235
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
946168-04-3
C14H17BO2
228.098
MFCD16294529
SJDFLZSEQGKSDJ-UHFFFAOYSA-N
2-3-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 3-ethynylbenzeneboronic acid pinacol ester, amtb210, 3-ethynylphenyl boronic acid pinacol ester, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2dioxaborolane
57416900
2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#C
This item is not returnable. View return policy
946168-04-3
C14H17BO2
228.098
MFCD16294529
SJDFLZSEQGKSDJ-UHFFFAOYSA-N
2-3-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 3-ethynylbenzeneboronic acid pinacol ester, amtb210, 3-ethynylphenyl boronic acid pinacol ester, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2dioxaborolane
57416900
2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 11140327
Formula Weight 228.1
Percent Purity 95%
Chemical Name or Material 3-Ethynylbenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'