missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexyl-1-propyne, 97%, Thermo Scientific Chemicals

Catalog Number p-7025156
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
17715-00-3
C9H14
122.211
MFCD00041562
UARFKZSJGDQRLF-UHFFFAOYSA-N
3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
87265
prop-2-ynylcyclohexane
C#CCC1CCCCC1
This item is not returnable. View return policy
17715-00-3
C9H14
122.211
MFCD00041562
UARFKZSJGDQRLF-UHFFFAOYSA-N
3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
87265
prop-2-ynylcyclohexane
C#CCC1CCCCC1

3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.845
Boiling Point 157°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.462
Quantity 1 g
UN Number UN3295
Beilstein 1849402
Solubility Information Slightly soluble in water.
Formula Weight 122.21
Percent Purity 97%
Chemical Name or Material 3-Cyclohexyl-1-propyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'