missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7054129
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
935685-88-4
C13H15BFNO2
247.08
MFCD11521351
WWDTYZLFIFQZDE-UHFFFAOYSA-N
3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-cyano-5-fluorobenzeneboronic acid pinacol ester, 3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-5-fluorophenylboronic acid pinacol ester, 3-fluoro-5-cyanophenylboronic acid pinacol ester, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3.2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46738194
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)C#N
This item is not returnable. View return policy
935685-88-4
C13H15BFNO2
247.08
MFCD11521351
WWDTYZLFIFQZDE-UHFFFAOYSA-N
3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-cyano-5-fluorobenzeneboronic acid pinacol ester, 3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-5-fluorophenylboronic acid pinacol ester, 3-fluoro-5-cyanophenylboronic acid pinacol ester, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3.2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46738194
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Cyano-5-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'