missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methoxyphenylboronic acid, 95%, Thermo Scientific™

Catalog Number p-3770820
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
175883-60-0
C7H8BClO3
186.40
MFCD04039888
VUTJHOWRPUPHIG-UHFFFAOYSA-N
3-chloro-4-methoxyphenyl boronic acid, 3-chloro-4-methoxybenzeneboronic acid, 4-borono-2-chloroanisole, 3-chloro-4-methoxyphenyl boranediol, boronic acid, 3-chloro-4-methoxyphenyl, akos brn-0653, 3-chloro-4, acmc-1c1io, ksc489k9h, 3-chloro-4-methoxyphenyl boronicacid
3247522
(3-chloro-4-methoxyphenyl)boronic acid
COC1=C(Cl)C=C(C=C1)B(O)O
This item is not returnable. View return policy
175883-60-0
C7H8BClO3
186.40
MFCD04039888
VUTJHOWRPUPHIG-UHFFFAOYSA-N
3-chloro-4-methoxyphenyl boronic acid, 3-chloro-4-methoxybenzeneboronic acid, 4-borono-2-chloroanisole, 3-chloro-4-methoxyphenyl boranediol, boronic acid, 3-chloro-4-methoxyphenyl, akos brn-0653, 3-chloro-4, acmc-1c1io, ksc489k9h, 3-chloro-4-methoxyphenyl boronicacid
3247522
(3-chloro-4-methoxyphenyl)boronic acid
COC1=C(Cl)C=C(C=C1)B(O)O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 240°C to 250°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(Cl)B(OH)2
Quantity 1 g
Formula Weight 186.4
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-methoxyphenylboronic acid
compliance-icons
Product Identifier
  • 3-Chloro-4-methoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'