missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7077636
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
930-96-1
C5H3BrOS
191.04
MFCD00126680
BCZHCWCOQDRYGS-UHFFFAOYSA-N
3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
2797079
3-bromothiophene-2-carbaldehyde
BrC1=C(SC=C1)C=O
This item is not returnable. View return policy
930-96-1
C5H3BrOS
191.04
MFCD00126680
BCZHCWCOQDRYGS-UHFFFAOYSA-N
3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
2797079
3-bromothiophene-2-carbaldehyde
BrC1=C(SC=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 24.0°C to 25.0°C
Density 1.8100g/mL
Boiling Point 92.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.81
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromothiophene-2-carboxaldehyde, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'