missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Catalog Number p-7032976
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1858240-14-8
C8H6BrClN2O3
293.501
MFCD22682824
RYTIKZUUZLHHDR-UHFFFAOYSA-N
3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate
121235573
3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid;hydrate
C1=CC2=NC(=C(N2C=C1Cl)Br)C(=O)O.O
This item is not returnable. View return policy
1858240-14-8
C8H6BrClN2O3
293.501
MFCD22682824
RYTIKZUUZLHHDR-UHFFFAOYSA-N
3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate
121235573
3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid;hydrate
C1=CC2=NC(=C(N2C=C1Cl)Br)C(=O)O.O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 216°C to 219°C (decomposition)
Quantity 250 mg
Formula Weight 275.49 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'