missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluoroaniline, 95%, Thermo Scientific Chemicals

Catalog Number 15417748
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
134168-97-1
C6H5BrFN
190.02
MFCD07779529
NGZAVSDIXFIWHJ-UHFFFAOYSA-N
3-fluoro-5-bromoaniline, 5-amino-1-bromo-3-fluorobenzene, 3-bromo-5-fluoro-phenylamine, benzenamine, 3-bromo-5-fluoro, 5-bromo-3-fluoroaniline, acmc-1bbb3, ksc120c3j, 3-bromo-5-fluorobenzenamine
15020155
3-bromo-5-fluoroaniline
NC1=CC(F)=CC(Br)=C1
This item is not returnable. View return policy
134168-97-1
C6H5BrFN
190.02
MFCD07779529
NGZAVSDIXFIWHJ-UHFFFAOYSA-N
3-fluoro-5-bromoaniline, 5-amino-1-bromo-3-fluorobenzene, 3-bromo-5-fluoro-phenylamine, benzenamine, 3-bromo-5-fluoro, 5-bromo-3-fluoroaniline, acmc-1bbb3, ksc120c3j, 3-bromo-5-fluorobenzenamine
15020155
3-bromo-5-fluoroaniline
NC1=CC(F)=CC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 250 mg
UN Number UN2810
Formula Weight 190.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluoroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'