missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluoro-5-nitropyridine, 97%

Product Code. 15448058
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15448058

Brand: Thermo Scientific Alfa Aesar H61500.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1868-58-2 | C5H2BrFN2O2 | 220.99 g/mol

3-Bromo-2-fluoro-5-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-fluoro-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1868-58-2
Molecular Formula C5H2BrFN2O2
Molecular Weight (g/mol) 220.99
MDL Number MFCD08703668
InChI Key NLLPMLSUMJHTCJ-UHFFFAOYSA-N
Synonym 2-fluoro-3-bromo-5-nitropyridine, acmc-1celf, 3-bromanyl-2-fluoranyl-5-nitro-pyridine
PubChem CID 345124
IUPAC Name 3-bromo-2-fluoro-5-nitropyridine
SMILES [O-][N+](=O)C1=CN=C(F)C(Br)=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 220.98
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluoro-5-nitropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.