missing translation for 'onlineSavingsMsg'
Learn More

3-(BOC-amino)pyrrolidine, 97%, Thermo Scientific™

Catalog Number p-4812856
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
99724-19-3
C9H18N2O2
186.26
DQQJBEAXSOOCPG-UHFFFAOYSA-N
3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine
2757234
tert-butyl N-pyrrolidin-3-ylcarbamate
CC(C)(C)OC(=O)NC1CCNC1
This item is not returnable. View return policy
99724-19-3
C9H18N2O2
186.26
DQQJBEAXSOOCPG-UHFFFAOYSA-N
3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine
2757234
tert-butyl N-pyrrolidin-3-ylcarbamate
CC(C)(C)OC(=O)NC1CCNC1
Melting Point 51°C to 54°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Formula Weight 186.26
Percent Purity 97%
Quantity 1 g
Chemical Name or Material 3-(BOC-amino)pyrrolidine
compliance-icons
Product Identifier
  • 3-Boc-aminopyrrolidine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'