Learn More
3-Amino-2,2-dimethyl-1-propanol, 97%, Thermo Scientific™
Catalog Number 15525246
Click to view available options
Quantity:
1 g
5 g
25 g
26734-09-8
C5H13NO
103.17
FNVOFDGAASRDQY-UHFFFAOYSA-N
3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
117326
3-amino-2,2-dimethylpropan-1-ol
CC(C)(CN)CO
26734-09-8
C5H13NO
103.17
FNVOFDGAASRDQY-UHFFFAOYSA-N
3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
117326
3-amino-2,2-dimethylpropan-1-ol
CC(C)(CN)CO
Melting Point 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 103.17
Percent Purity 97%
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol
compliance-icons
Product Identifier
  • 3-Amino-2,2-dimethyl-1-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.