missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromobenzotrifluoride, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,5-Dibromobenzotrifluoride used as a intermediate in organic synthesis and pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 4°C. Storage: Keep container tightly closed. Store in a cool, dry and well-ventilated place.
3,5-Dibromobenzotrifluoride used as a intermediate in organic synthesis and pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 4°C. Storage: Keep container tightly closed. Store in a cool, dry and well-ventilated place.
Specifications
Color | Yellow |
Quantity | 25 g |
Beilstein | 2093912 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 303.9 |
Percent Purity | 96% |
Physical Form | Liquid |
Chemical Name or Material | 3,5-Dibromobenzotrifluoride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'