Learn More
3-(4-Methylpiperazin-1-yl)aniline, 97%, Thermo Scientific™
Catalog Number p-4510561
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
148546-99-0
C11H17N3
191.278
RJGHJWKQCJAJEP-UHFFFAOYSA-N
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
11564613
3-(4-methylpiperazin-1-yl)aniline
CN1CCN(CC1)C2=CC(=CC=C2)N
148546-99-0
C11H17N3
191.278
RJGHJWKQCJAJEP-UHFFFAOYSA-N
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
11564613
3-(4-methylpiperazin-1-yl)aniline
CN1CCN(CC1)C2=CC(=CC=C2)N
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 3-(4-Methylpiperazin-1-yl)aniline