Learn More
3,4,5-Trichloropyridine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 72.0°C to 73.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Formula Weight | 182.44 |
Percent Purity | 98% |
Chemical Name or Material | 3,4,5-Trichloropyridine |