Learn More
3,3-Dimethylbutyraldehyde, Thermo Scientific Chemicals
Catalog Number p-4089664
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
2987-16-8
C6H12O
100.16
MFCD00042807
LTNUSYNQZJZUSY-UHFFFAOYSA-N
3,3-dimethylbutyraldehyde, 3,3-dimethyl-butyraldehyde, butanal, 3,3-dimethyl, unii-cam6hd7jki, tert-butylacetaldehyde, 3,3-dimethyl-butanal, cam6hd7jki, neohexal, pubchem3076, 3,3-dimethylbutyraldehyd
76335
3,3-dimethylbutanal
CC(C)(C)CC=O
2987-16-8
C6H12O
100.16
MFCD00042807
LTNUSYNQZJZUSY-UHFFFAOYSA-N
3,3-dimethylbutyraldehyde, 3,3-dimethyl-butyraldehyde, butanal, 3,3-dimethyl, unii-cam6hd7jki, tert-butylacetaldehyde, 3,3-dimethyl-butanal, cam6hd7jki, neohexal, pubchem3076, 3,3-dimethylbutyraldehyd
76335
3,3-dimethylbutanal
CC(C)(C)CC=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -24°C
Color Colorless
Density 0.7980g/mL
Boiling Point 104°C to 106°C
Flash Point 13°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)3CCH2CHO
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.798
Solubility Information Solubility in water: 7.3g/L (20°C)
Viscosity 0.98 mPa.s (20°C)
Formula Weight 100.16
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylbutyraldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethylbutyraldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.