missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals

Catalog Number p-7021610
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
917-92-0
C6H10
82.146
MFCD00008852
PPWNCLVNXGCGAF-UHFFFAOYSA-N
13512
3,3-dimethylbut-1-yne
CC(C)(C)C#C
This item is not returnable. View return policy
917-92-0
C6H10
82.146
MFCD00008852
PPWNCLVNXGCGAF-UHFFFAOYSA-N
13512
3,3-dimethylbut-1-yne
CC(C)(C)C#C

3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

Solubility
Miscible with chloroform, benzene and toluene. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -78°C
Density 0.671
Boiling Point 37°C to 38°C
Flash Point <−34°C (−29°F)
Odor Recognizable
Linear Formula (CH3)3CC≡CH
Refractive Index 1.3745
Quantity 5 g
UN Number UN3295
Beilstein 1697100
Synonym 3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
Solubility Information Miscible with chloroform,benzene and toluene. Immiscible with water.
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 3,3-Dimethyl-1-butyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'