Learn More
(2RS)-2-(1-Methylethyl)pentanoic Acid, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
(2RS)-2-(1-Methylethyl)pentanoic Acid
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(1-Methylethyl)pentanoicAcid |
| CAS | 62391-99-5 |
| Analyte or Component Names | (2RS)-2-(1-Methylethyl)pentanoicAcid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H16 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Valproic Acid Imp. C (EP), Valproic Acid USP Related Compound B, Valproic Acid USP RC B, (2RS)-2-(1-Methylethyl)pentanoicAcid, Valproic Acid Related Compound B, Valproic Acid Impurity C |
| InChI Formula | InChI=1S/C8H16O2/c1-4-5-7(6(2)3)8(9)10/h6-7H,4-5H2,1-3H3,(H,9,10) |
| SMILES | CCCC(C(C)C)C(=O)O |
| IUPAC Name | 2-propan-2-ylpentanoic acid |
| Show More |