missing translation for 'onlineSavingsMsg'
Learn More

(2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one, 95%, Thermo Scientific™

Catalog Number p-7031706
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
14575-25-8
C21H21NO
303.405
MFCD06409497
FWAPKZOTGYVZSA-UHFFFAOYSA-N
e-1-phenyl-4-e-1,3,3-trimethylindolin-2-ylidene but-2-en-1-one, 2-buten-1-one,4-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-1-phenyl, 1-phenyl-4-1,3,3-trimethylindol-2-ylidene but-2-en-1-one, 1-phenyl-4-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene but-2-en-1-one
53427010
1-phenyl-4-(1,3,3-trimethylindol-2-ylidene)but-2-en-1-one
CC1(C2=CC=CC=C2N(C1=CC=CC(=O)C3=CC=CC=C3)C)C
This item is not returnable. View return policy
14575-25-8
C21H21NO
303.405
MFCD06409497
FWAPKZOTGYVZSA-UHFFFAOYSA-N
e-1-phenyl-4-e-1,3,3-trimethylindolin-2-ylidene but-2-en-1-one, 2-buten-1-one,4-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-1-phenyl, 1-phenyl-4-1,3,3-trimethylindol-2-ylidene but-2-en-1-one, 1-phenyl-4-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene but-2-en-1-one
53427010
1-phenyl-4-(1,3,3-trimethylindol-2-ylidene)but-2-en-1-one
CC1(C2=CC=CC=C2N(C1=CC=CC(=O)C3=CC=CC=C3)C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 162°C
Quantity 250 mg
Formula Weight 303.41
Percent Purity 95%
Chemical Name or Material (2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'