Learn More
Thermo Scientific™ 2000mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
Click to view available options
Sterility:
Non-sterile
Sterilized using Beta irradiation following standards ISO 11137-1 and ISO 11137-2 (non-sterile)
Description
Process a 16L harvest in a single run with the Thermo Scientific™ 2000 mL Bottle, the ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
- Ideal for bacterial, yeast, mamalian cell separations
- Process up to 12L in a single run at speeds up to 7,187 x g or 16L in a single run at speeds up to 5,374 x g
- Wide-mouth design simplifies decanting, re-suspensions and pellet removal
- For centrifugation operating temperatures from 4° to 40°C
Specifications
Capacity | 2000 mL |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
For Use With (Equipment) | BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swinging Bucket Rotor |
Through the Closure Dispensing | No |
Product Type | Bottle |
Capacity (Metric) | 2000 mL |
Capacity (English) | 67.62 oz. |
Color | White |
Description | 2000 mL bottle |
Show More |