missing translation for 'onlineSavingsMsg'
Learn More

2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%, Thermo Scientific Chemicals

Catalog Number 15428316
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
315-08-2
C10H6F3NOS2
277.28
MFCD00126176
MRDJXDJVVCNDND-UHFFFAOYSA-N
n-3-trifluoromethyl phenyl rhodanine, 2-thioxo-3-3-trifluoromethyl-phenyl-thiazolidin-4-one, 2-sulfanylidene-3-3-trifluoromethyl phenyl-1,3-thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl-4-thiazolidinone, 2-thioxo-3-3-trifluoromethylhenyl-thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl-1,3-thiazolidin-4-one, 3-3-trifluoromethyl phenyl rhodanine, 2-tioxo-3-3-trifluoromethylphenyl thiazolidin-4-one
278104
2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
FC(F)(F)C1=CC=CC(=C1)N1C(=O)CSC1=S
This item is not returnable. View return policy
315-08-2
C10H6F3NOS2
277.28
MFCD00126176
MRDJXDJVVCNDND-UHFFFAOYSA-N
n-3-trifluoromethyl phenyl rhodanine, 2-thioxo-3-3-trifluoromethyl-phenyl-thiazolidin-4-one, 2-sulfanylidene-3-3-trifluoromethyl phenyl-1,3-thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl-4-thiazolidinone, 2-thioxo-3-3-trifluoromethylhenyl-thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl-1,3-thiazolidin-4-one, 3-3-trifluoromethyl phenyl rhodanine, 2-tioxo-3-3-trifluoromethylphenyl thiazolidin-4-one
278104
2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
FC(F)(F)C1=CC=CC(=C1)N1C(=O)CSC1=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Formula Weight 277.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'