missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrimidinylthiourea, 97+%, Thermo Scientific Chemicals

Catalog Number p-7032282
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
31437-20-4
C5H6N4S
154.191
MFCD02662218
MCCLFFQZNBEOAV-UHFFFAOYSA-N
1-pyrimidin-2-yl thiourea, n-pyrimidin-2-ylthiourea, pyrimidin-2-yl-thiourea, pyrimidin-2-yl thiourea, n-2-pyrimidinyl thiourea, n-pyrimidin-2ylthiourea, 1-pyrimidin-2-ylthiourea, 1-pyrimidin-2-yl-thiourea, amino pyrimidin-2-ylamino methane-1-thione
2760516
pyrimidin-2-ylthiourea
C1=CN=C(N=C1)NC(=S)N
This item is not returnable. View return policy
31437-20-4
C5H6N4S
154.191
MFCD02662218
MCCLFFQZNBEOAV-UHFFFAOYSA-N
1-pyrimidin-2-yl thiourea, n-pyrimidin-2-ylthiourea, pyrimidin-2-yl-thiourea, pyrimidin-2-yl thiourea, n-2-pyrimidinyl thiourea, n-pyrimidin-2ylthiourea, 1-pyrimidin-2-ylthiourea, 1-pyrimidin-2-yl-thiourea, amino pyrimidin-2-ylamino methane-1-thione
2760516
pyrimidin-2-ylthiourea
C1=CN=C(N=C1)NC(=S)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 234°C to 238°C
Quantity 1 g
Formula Weight 154.19
Percent Purity ≥97%
Chemical Name or Material 2-Pyrimidinylthiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'